Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Serping1
Entrez Gene ID
12258
Gene Name
serine (or cysteine) peptidase inhibitor, clade G, member 1
Gene Aliases
C1INH, C1nh
Chromosome Location
Chr.2: 84765360 - 84775445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009776.3 NP_033906.2 2 - 3 137 99
GenBank mRNA AF010254.1 - 2 - 3 112 99
AK144292.1 - 1 - 2 98 99
AK165744.1 - 1 - 2 85 99
AK167585.1 - 1 - 2 75 99
BC002026.1 - 1 - 2 68 99
CA458230.1 - 2 - 3 174 99
Y10386.1 - 2 - 3 94 99

View Full Product Details