Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdsub1
Entrez Gene ID
72137
Gene Name
WD repeat, SAM and U-box domain containing 1
Gene Aliases
1700048E19Rik, 2610014F08Rik
Chromosome Location
Chr.2: 59852364 - 59882625 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159636.1 NP_001153108.1 4 - 5 897 80
NM_028118.2 NP_082394.1 4 - 5 897 80
GenBank mRNA AK011391.1 - 4 - 5 833 80
BC050792.1 - 4 - 5 838 80
BC061948.1 - 4 - 5 875 80

View Full Product Details