Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hdgfrp2
Entrez Gene ID
15193
Gene Name
hepatoma-derived growth factor, related protein 2
Gene Aliases
HRP-2
Chromosome Location
Chr.17: 56079604 - 56100601 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK032572.1 - 5 - 6 796 62
AK143616.1 - 5 - 6 717 62
AK146813.1 - 5 - 6 656 62
AK146918.1 - 5 - 6 694 62
BC003741.1 - 5 - 6 653 62
D63850.1 - 5 - 6 697 62
FN687734.2 - 5 - 6 440 62

View Full Product Details