Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spats1
Entrez Gene ID
71020
Gene Name
spermatogenesis associated, serine-rich 1
Gene Aliases
1700011H05Rik, 4933400B06Rik, Daip2, Ddip, Srsp1
Chromosome Location
Chr.17: 45445877 - 45475194 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027649.3 NP_081925.2 4 - 5 609 77
GenBank mRNA AK005865.1 - 4 - 5 610 77
AK016586.1 - 4 - 5 536 77
AK078278.1 - - 531 77
AY336501.1 - 4 - 5 497 77
BC131909.1 - 4 - 5 569 77

View Full Product Details