The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011917.2 | NP_036047.2 | 1 - 2 | 140 | 76 |
GenBank mRNA | AK004800.1 | - | 1 - 2 | 115 | 76 |
AK011711.1 | - | 1 - 2 | 138 | 76 | |
AK031247.1 | - | 1 - 2 | 183 | 76 | |
AK053643.1 | - | 1 - 2 | 133 | 76 | |
AK133654.1 | - | 1 - 2 | 122 | 76 | |
AK163899.1 | - | 1 - 2 | 118 | 76 | |
AK167019.1 | - | 1 - 2 | 160 | 76 | |
AK168037.1 | - | 1 - 2 | 186 | 76 | |
BC052395.1 | - | 1 - 2 | 116 | 76 | |
BC052823.1 | - | 1 - 2 | 105 | 76 | |
BC054743.1 | - | 1 - 2 | 140 | 76 | |
D38517.1 | - | 1 - 2 | 78 | 76 |