Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rin2
- Entrez Gene ID
-
74030
- Gene Name
- Ras and Rab interactor 2
- Gene Aliases
-
2010003K16Rik, 4632403N06Rik, AW821980, RASSF4
- Chromosome Location
-
Chr.2: 145771727 - 145887616 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CN461821.1 |
- |
3 - 4 |
393 |
58 |
View Full Product Details