The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144804.1 | NP_659053.1 | 8 - 9 | 1391 | 132 |
GenBank mRNA | AK134830.1 | - | 8 - 9 | 1411 | 132 |
AK144066.1 | - | 8 - 9 | 1404 | 132 | |
AK162086.1 | - | 8 - 9 | 1415 | 132 | |
AK162218.1 | - | 8 - 9 | 1414 | 132 | |
BC013499.1 | - | 8 - 9 | 1391 | 132 |