The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033235.3 | NP_001028407.1 | 3 - 4 | 1090 | 79 |
NM_001172168.1 | NP_001165639.1 | 3 - 4 | 1090 | 79 | |
GenBank mRNA | AK136498.1 | - | 3 - 4 | 1010 | 79 |
BC147336.1 | - | 2 - 3 | 781 | 79 | |
BC147337.1 | - | 2 - 3 | 781 | 79 | |
BC172037.1 | - | 2 - 3 | 781 | 79 |