Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Folr2
- Entrez Gene ID
-
14276
- Gene Name
- folate receptor 2 (fetal)
- Gene Aliases
-
FBP2, FR-P3, FR-beta, Folbp-2, Folbp2
- Chromosome Location
-
Chr.7: 101839988 - 101857190 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
M64817.1 |
- |
2 - 3 |
264 |
71 |
View Full Product Details