Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bub1
Entrez Gene ID
12235
Gene Name
budding uninhibited by benzimidazoles 1 homolog (S. cerevisiae)
Gene Aliases
AL022991, Bub1a, C80208, D2Xrf87
Chromosome Location
Chr.2: 127800200 - 127831876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113179.1 NP_001106650.1 6 - 7 688 138
NM_009772.2 NP_033902.2 6 - 7 688 138
GenBank mRNA AF002823.1 - 6 - 7 634 138
AK021258.1 - 6 - 7 679 138
AK042932.1 - 6 - 7 689 138
AK144213.1 - 6 - 7 697 138
BC029116.1 - 6 - 7 654 138
U89795.1 - 6 - 7 706 138

View Full Product Details