The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010403.2 | NP_034533.1 | 7 - 8 | 1059 | 101 |
GenBank mRNA | AF104312.1 | - | 7 - 8 | 1059 | 101 |
AK149565.1 | - | 7 - 8 | 1066 | 101 | |
BC018587.1 | - | 7 - 8 | 1037 | 101 | |
BC119535.1 | - | 7 - 8 | 1059 | 101 | |
BC119536.1 | - | 7 - 8 | 1059 | 101 |