Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nuf2
Entrez Gene ID
66977
Gene Name
NUF2, NDC80 kinetochore complex component, homolog (S. cerevisiae)
Gene Aliases
2410003C07Rik, AU044112, C85691, Cdca1, NUF2R
Chromosome Location
Chr.1: 169497934 - 169531464 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023284.3 NP_075773.2 3 - 4 362 79
GenBank mRNA AF326732.1 - 4 - 5 403 79
AK010351.1 - 3 - 4 284 79
AK012011.1 - 3 - 4 296 79
AK132467.1 - 3 - 4 363 79
AK162249.1 - 4 - 5 398 79
BC020026.1 - 4 - 5 371 79

View Full Product Details