Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fxr1
Entrez Gene ID
14359
Gene Name
fragile X mental retardation gene 1, autosomal homolog
Gene Aliases
1110050J02Rik, 9530073J07Rik, AI851072, Fxr1h, Fxr1p
Chromosome Location
Chr.3: 34020058 - 34069980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113188.1 NP_001106659.1 1 - 2 83 63
NM_001113189.1 NP_001106660.1 1 - 2 83 63
NM_008053.2 NP_032079.1 1 - 2 83 63
GenBank mRNA BC019139.1 - 1 - 2 64 63
CJ086949.1 - 1 - 2 84 63
X90875.1 - 1 - 2 62 63

View Full Product Details