Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Prrx1
- Entrez Gene ID
-
18933
- Gene Name
- paired related homeobox 1
- Gene Aliases
-
A230024N07Rik, AA755424, AI385634, AI843499, K-2, Pmx, Pmx1, Prx1, mHox
- Chromosome Location
-
Chr.1: 163245119 - 163313705 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK008428.2 |
- |
2 - 3 |
81 |
97 |
View Full Product Details