Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fnip1
Entrez Gene ID
216742
Gene Name
folliculin interacting protein 1
Gene Aliases
A730024A03Rik, AI838773, AW557298
Chromosome Location
Chr.11: 54438146 - 54518241 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173753.4 NP_776114.2 10 - 11 1236 93
GenBank mRNA AK087849.1 - 10 - 11 1890 93
AK122574.1 - 10 - 11 1184 93
BC079892.1 - 10 - 11 1226 93

View Full Product Details