Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dppa1
Entrez Gene ID
347708
Gene Name
developmental pluripotency associated 1
Gene Aliases
C77727
Chromosome Location
Chr.11: 46607010 - 46632233 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178247.3 NP_839978.1 1 - 2 43 78
GenBank mRNA AK145642.1 - 1 - 2 44 78
AK166752.1 - 1 - 2 44 78

View Full Product Details