The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145919.1 | NP_001139391.1 | 2 - 3 | 366 | 107 |
NM_172677.3 | NP_766265.3 | 2 - 3 | 366 | 107 | |
GenBank mRNA | AK137245.1 | - | 2 - 3 | 313 | 107 |
BC067040.1 | - | 2 - 3 | 337 | 107 | |
BC067042.1 | - | 2 - 3 | 299 | 107 |