The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145919.1 | NP_001139391.1 | 3 - 4 | 442 | 151 |
GenBank mRNA | BC067040.1 | - | 3 - 4 | 413 | 151 |