Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pld1
Entrez Gene ID
18805
Gene Name
phospholipase D1
Gene Aliases
AA536939, C85393, Pld1a, Pld1b
Chromosome Location
Chr.3: 27938680 - 28133362 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164056.1 NP_001157528.1 19 - 20 2366 112
NM_008875.4 NP_032901.2 18 - 19 2311 112
GenBank mRNA AK136111.1 - 13 - 14 1549 112
BC068144.1 - 18 - 19 2168 112
U87868.1 - 19 - 20 2349 112

View Full Product Details