The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164056.1 | NP_001157528.1 | 8 - 9 | 916 | 94 |
NM_008875.4 | NP_032901.2 | 7 - 8 | 861 | 94 | |
GenBank mRNA | AK136111.1 | - | 2 - 3 | 99 | 94 |
BC068144.1 | - | 7 - 8 | 718 | 94 | |
U87868.1 | - | 8 - 9 | 899 | 94 |