The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033409.3 | NP_001028581.1 | 16 - 17 | 1704 | 66 |
GenBank mRNA | AK052873.1 | - | 11 - 12 | 904 | 66 |
AK085901.1 | - | 16 - 17 | 1663 | 66 | |
AK137380.1 | - | 16 - 17 | 1705 | 66 | |
BC026896.1 | - | 1 - 2 | 46 | 66 | |
BC141210.1 | - | 16 - 17 | 1656 | 66 |