Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ispd
Entrez Gene ID
75847
Gene Name
isoprenoid synthase domain containing
Gene Aliases
4930579E17Rik, AV040780
Chromosome Location
Chr.12: 36380585 - 36689503 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289503.1 NP_001276432.1 1 - 2 155 80
GenBank mRNA AK016317.1 - 1 - 2 123 80

View Full Product Details