Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sav1
Entrez Gene ID
64010
Gene Name
salvador homolog 1 (Drosophila)
Gene Aliases
1700040G09Rik, Salv, Sav, WW45, Wwp3, Wwp4
Chromosome Location
Chr.12: 69965012 - 69987002 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022028.2 NP_071311.1 2 - 3 773 69
GenBank mRNA AJ292968.1 - 2 - 3 773 69
AK006655.1 - 1 - 2 210 69
BC019377.1 - 2 - 3 706 69

View Full Product Details