Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gli2
Entrez Gene ID
14633
Gene Name
GLI-Kruppel family member GLI2
Gene Aliases
AW546128
Chromosome Location
Chr.1: 118834061 - 119053619 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081125.1 NP_001074594.1 9 - 10 1657 65
GenBank mRNA BC082604.1 - 9 - 10 1598 65
BC085190.1 - 9 - 10 1573 65
X99104.1 - 9 - 10 1417 65

View Full Product Details