Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfcp2l1
Entrez Gene ID
81879
Gene Name
transcription factor CP2-like 1
Gene Aliases
1810030F05Rik, 4932442M07Rik, AA575098, Cp2l1, Crtr-1, D930018N21Rik, LBP-9, Tcfcp2l1
Chromosome Location
Chr.1: 118627923 - 118685168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023755.2 NP_076244.2 7 - 8 864 76
GenBank mRNA AK143960.1 - 7 - 8 863 76
BC116663.1 - 7 - 8 859 76

View Full Product Details