The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011439.2 | NP_035569.2 | 13 - 14 | 1910 | 75 |
GenBank mRNA | AB006329.1 | - | 12 - 13 | 1566 | 75 |
BC007130.1 | - | 13 - 14 | 1732 | 75 |