Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epn3
Entrez Gene ID
71889
Gene Name
epsin 3
Gene Aliases
2310022G12Rik
Chromosome Location
Chr.11: 94489599 - 94499974 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027984.3 NP_082260.1 3 - 4 1079 71
GenBank mRNA AK009469.1 - 2 - 3 459 71
BC016454.1 - 3 - 4 1028 71

View Full Product Details