Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap23
Entrez Gene ID
58996
Gene Name
Rho GTPase activating protein 23
Gene Aliases
A330041B18Rik
Chromosome Location
Chr.11: 97404393 - 97502402 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB041572.1 - 3 - 4 165 59

View Full Product Details