The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177324.1 | NP_001170795.1 | 1 - 2 | 321 | 78 |
NM_001253706.1 | NP_001240635.1 | 1 - 2 | 321 | 78 | |
NM_019942.5 | NP_064326.2 | 1 - 2 | 321 | 78 | |
GenBank mRNA | AB023622.1 | - | 1 - 2 | 85 | 78 |
AK083302.1 | - | 1 - 2 | 121 | 78 | |
AK088406.1 | - | 1 - 2 | 101 | 78 | |
AK088614.1 | - | 1 - 2 | 84 | 78 | |
AK160884.1 | - | 1 - 2 | 115 | 78 | |
AK162755.1 | - | 1 - 2 | 121 | 78 | |
AK172894.1 | - | 1 - 2 | 102 | 78 | |
BC010489.1 | - | 1 - 2 | 107 | 78 | |
BY776992.1 | - | 1 - 2 | 321 | 78 |