The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040395.4 | NP_001035485.2 | 3 - 4 | 679 | 62 |
NM_001085410.2 | NP_001078879.1 | 3 - 4 | 679 | 62 | |
NM_001286253.1 | NP_001273182.1 | 3 - 4 | 679 | 62 | |
NM_001286255.1 | NP_001273184.1 | 3 - 4 | 679 | 62 | |
GenBank mRNA | AK003858.1 | - | 3 - 4 | 256 | 62 |
AK016988.1 | - | 3 - 4 | 245 | 62 | |
AK046666.1 | - | 3 - 4 | 680 | 62 | |
AK078318.1 | - | 3 - 4 | 282 | 62 | |
AK149267.1 | - | 3 - 4 | 259 | 62 | |
AK162601.1 | - | 3 - 4 | 253 | 62 | |
BC115776.1 | - | 3 - 4 | 258 | 62 |