The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027644.1 | NP_081920.1 | 6 - 7 | 748 | 64 |
GenBank mRNA | AB049453.1 | - | 6 - 7 | 748 | 64 |
AK016509.1 | - | 6 - 7 | 726 | 64 | |
BC145586.1 | - | 6 - 7 | 783 | 64 | |
BC150797.1 | - | 6 - 7 | 764 | 64 |