Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xpnpep3
Entrez Gene ID
321003
Gene Name
X-prolyl aminopeptidase (aminopeptidase P) 3, putative
Gene Aliases
APP3, E430012M05Rik
Chromosome Location
Chr.15: 81400128 - 81457476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK042110.1 - 2 - 3 258 71
AK158394.1 - 2 - 3 239 71

View Full Product Details