The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001321.3 | NP_001001321.2 | 9 - 10 | 742 | 100 |
GenBank mRNA | AB117931.1 | - | 9 - 10 | 723 | 100 |
AB117932.1 | - | 9 - 10 | 723 | 100 | |
AK154257.1 | - | 9 - 10 | 768 | 100 | |
AK159148.1 | - | 9 - 10 | 743 | 100 | |
BC046402.1 | - | 3 - 4 | 282 | 100 | |
BC107210.1 | - | 8 - 9 | 689 | 100 | |
BC107211.1 | - | 9 - 10 | 722 | 100 |