Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdk12
Entrez Gene ID
69131
Gene Name
cyclin-dependent kinase 12
Gene Aliases
1810022J16Rik, AI646528, Crk7, Crkrs, D11Ertd752e, Pksc
Chromosome Location
Chr.11: 98202326 - 98281101 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109626.1 NP_001103096.1 9 - 10 2899 99
NM_001109628.1 NP_001103098.1 9 - 10 2899 99
NM_026952.2 NP_081228.2 9 - 10 2899 99
GenBank mRNA AK049376.1 - 9 - 10 2545 99
AK129237.1 - 9 - 10 2038 99
AK137734.1 - 7 - 8 769 99
AY072295.1 - 9 - 10 2890 99
BC116645.1 - 9 - 10 2882 99

View Full Product Details