Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfr
Entrez Gene ID
22763
Gene Name
zinc finger RNA binding protein
Gene Aliases
C920030H05Rik
Chromosome Location
Chr.15: 12117793 - 12185449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011767.2 NP_035897.2 9 - 10 1772 115
GenBank mRNA AF071059.1 - 9 - 10 1770 115
AK034963.1 - 2 - 3 341 115
AK158938.1 - 1 - 2 80 115
BC094220.1 - 8 - 9 1659 115
BC141139.1 - 9 - 10 1756 115

View Full Product Details