Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sele
Entrez Gene ID
20339
Gene Name
selectin, endothelial cell
Gene Aliases
CD62E, E-selectin, Elam
Chromosome Location
Chr.1: 164039604 - 164058487 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011345.2 NP_035475.1 8 - 9 1415 86
GenBank mRNA AK153623.1 - 8 - 9 1434 86
BC119083.1 - 8 - 9 1366 86
BC120795.1 - 8 - 9 1366 86
M87862.1 - 8 - 9 1415 86

View Full Product Details