Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rb1
Entrez Gene ID
19645
Gene Name
retinoblastoma 1
Gene Aliases
Rb, Rb-1, pRb
Chromosome Location
Chr.14: 73194453 - 73325887 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009029.2 NP_033055.2 23 - 24 2619 76
GenBank mRNA AK132875.1 - 12 - 13 1365 76
AK141028.1 - 23 - 24 2651 76
AK146467.1 - 18 - 19 1950 76
BC096525.1 - 23 - 24 2619 76
DQ400415.1 - 23 - 24 2469 76
M26391.1 - 23 - 24 2567 76

View Full Product Details