The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007446.2 | NP_031472.2 | 1 - 2 | 148 | 89 |
GenBank mRNA | AK029642.1 | - | 1 - 2 | 71 | 89 |
CK791844.1 | - | 1 - 2 | 81 | 89 | |
V00719.1 | - | 1 - 2 | 148 | 89 |