Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a1
Entrez Gene ID
20517
Gene Name
solute carrier family 22 (organic cation transporter), member 1
Gene Aliases
Lx1, Oct1, Orct, Orct1
Chromosome Location
Chr.17: 12648874 - 12675838 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009202.5 NP_033228.2 4 - 5 1020 86
GenBank mRNA AF010259.1 - 4 - 5 878 86
BC021651.1 - 4 - 5 1020 86
U38652.1 - 4 - 5 875 86

View Full Product Details