Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pspc1
Entrez Gene ID
66645
Gene Name
paraspeckle protein 1
Gene Aliases
5730470C09Rik, AI327109, AI449052, PSP1
Chromosome Location
Chr.14: 56722441 - 56778316 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025682.3 NP_079958.3 6 - 7 1319 67
GenBank mRNA AK011272.1 - 6 - 7 1316 67
AK017689.1 - 6 - 7 1320 67
AK150823.1 - 6 - 7 1320 67
AK160722.1 - 6 - 7 1320 67
BC026772.1 - 6 - 7 1298 67

View Full Product Details