Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Osgep
Entrez Gene ID
66246
Gene Name
O-sialoglycoprotein endopeptidase
Gene Aliases
1500019L24Rik, GCPL-1, PRSMG1
Chromosome Location
Chr.14: 50915374 - 50924893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133676.2 NP_598437.2 6 - 7 752 60
GenBank mRNA AK005291.1 - 6 - 7 703 60
AK049955.1 - - 693 60
AK076246.1 - 6 - 7 712 60
AK088346.1 - 6 - 7 715 60
AK142730.1 - 6 - 7 752 60
AK145181.1 - 6 - 7 680 60
AK157554.1 - - 703 60
AK166415.1 - 6 - 7 712 60
AK167893.1 - 6 - 7 694 60
BC002296.1 - 6 - 7 687 60
BC091757.1 - 6 - 7 674 60

View Full Product Details