The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031367.1 | NP_112735.1 | - | 73 | 62 |
GenBank mRNA | AF336221.1 | - | - | 73 | 62 |
BC024930.1 | - | - | 103 | 62 |