The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163457.1 | NP_001156929.1 | 7 - 8 | 72 | |
NM_008642.2 | NP_032668.2 | 5 - 6 | 72 | ||
GenBank mRNA | AK136278.1 | - | 7 - 8 | 72 | |
AK146553.1 | - | 5 - 6 | 72 | ||
BC012686.1 | - | 5 - 6 | 653 | 72 | |
EU553486.1 | - | 5 - 6 | 661 | 72 | |
L47970.1 | - | 5 - 6 | 640 | 72 |