Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcf11
Entrez Gene ID
74737
Gene Name
cleavage and polyadenylation factor subunit homolog (S. cerevisiae)
Gene Aliases
2500001H09Rik, 5730417B17Rik, C77803
Chromosome Location
Chr.7: 92643458 - 92670087 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029078.3 NP_083354.3 3 - 4 797 94
GenBank mRNA AK161260.1 - 3 - 4 821 94
AK161614.1 - 3 - 4 787 94
AK173035.1 - 3 - 4 769 94
BC150781.1 - 3 - 4 633 94

View Full Product Details