Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap29
Entrez Gene ID
214137
Gene Name
Rho GTPase activating protein 29
Gene Aliases
6720461J18Rik, A830014I19, AU040217, B130017I01Rik, C76601, Parg1
Chromosome Location
Chr.3: 121953072 - 122016157 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172525.2 NP_766113.1 16 - 17 1898 107
GenBank mRNA AK043637.1 - 16 - 17 1945 107
AK044979.1 - 16 - 17 1948 107
AK132716.1 - 11 - 12 1284 107
BC031572.1 - 5 - 6 565 107
BC040387.1 - 16 - 17 1922 107

View Full Product Details