Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arhgef15
Entrez Gene ID
442801
Gene Name
Rho guanine nucleotide exchange factor (GEF) 15
Gene Aliases
D130071N09, D530030K12Rik, E5
Chromosome Location
Chr.11: 68927611 - 68956934 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177566.3 NP_808234.2 2 - 3 1024 95
GenBank mRNA BC089365.1 - 2 - 3 1024 95
BC116343.1 - 2 - 3 1000 95
BC116344.1 - 2 - 3 1000 95

View Full Product Details