Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr76
Entrez Gene ID
241627
Gene Name
WD repeat domain 76
Gene Aliases
5830411K18Rik, 6030457D09, AA517836
Chromosome Location
Chr.2: 121506723 - 121544860 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290986.1 NP_001277915.1 10 - 11 1462 72
NM_001290987.1 NP_001277916.1 9 - 10 1347 72
NM_030234.2 NP_084510.2 10 - 11 1477 72
GenBank mRNA AK031592.1 - 10 - 11 1403 72
AK162181.1 - 10 - 11 1430 72
BC094676.1 - 9 - 10 1113 72
BC117821.1 - 9 - 10 1286 72
BC117822.1 - 10 - 11 1416 72

View Full Product Details