The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145900.1 | NP_001139372.1 | 1 - 2 | 27 | 116 |
GenBank mRNA | AI119004.1 | - | 1 - 2 | 27 | 116 |