The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145900.1 | NP_001139372.1 | 2 - 3 | 344 | 67 |
NM_201646.2 | NP_964008.2 | 1 - 2 | 426 | 67 | |
GenBank mRNA | AI119004.1 | - | 2 - 3 | 344 | 67 |
AK146991.1 | - | 1 - 2 | 258 | 67 | |
BC031195.1 | - | 1 - 2 | 426 | 67 |