The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204165.1 | NP_001191094.1 | 10 - 11 | 1081 | 84 |
NM_001204166.1 | NP_001191095.1 | 7 - 8 | 973 | 84 | |
NM_019688.2 | NP_062662.1 | 9 - 10 | 1027 | 84 | |
GenBank mRNA | AB021132.1 | - | 10 - 11 | 1056 | 84 |
AB037668.1 | - | 1 - 2 | 416 | 84 | |
AB445011.1 | - | 7 - 8 | 965 | 84 | |
AF115480.1 | - | 9 - 10 | 955 | 84 | |
AK004874.1 | - | 1 - 2 | 73 | 84 | |
AK220281.1 | - | 10 - 11 | 1081 | 84 | |
BC139822.1 | - | 10 - 11 | 1013 | 84 |